missing translation for 'onlineSavingsMsg'
Learn More

Silver perchlorate monohydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048836
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
This item is not returnable. View return policy
14242-05-8
AgClO4
207.31
MFCD00149128
YDHABVNRCBNRNZ-UHFFFAOYSA-M
silver perchlorate hydrate, silver perchlorate monohydrate, silver perchlorite, silver perchloric acid, acmc-1brvv, ag.clo4.h2o, silver i perchlorate monohydrate, silver 1+ ion hydrate perchlorate, silver 1+ hydrate perchlorate
15959344
silver;perchlorate;hydrate
[Ag+].[O-][Cl](=O)(=O)=O
This item is not returnable. View return policy
14242-05-8
AgClO4
207.31
MFCD00149128
YDHABVNRCBNRNZ-UHFFFAOYSA-M
silver perchlorate hydrate, silver perchlorate monohydrate, silver perchlorite, silver perchloric acid, acmc-1brvv, ag.clo4.h2o, silver i perchlorate monohydrate, silver 1+ ion hydrate perchlorate, silver 1+ hydrate perchlorate
15959344
silver;perchlorate;hydrate
[Ag+].[O-][Cl](=O)(=O)=O

Silver perchlorate can be used in the manufacturing of explosives. It is generally used as a source of silver ions during silver nanoparticles synthesis. It is used as a catalyst in organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 485°C (decomposition)
Color White
Physical Form Crystalline
Quantity 2 g
UN Number UN1481
Merck Index 14,8523
Solubility Information Soluble in water and organic solvents.
Formula Weight 225.34 (207.32 Anhydrous)
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 2.800 g/mL
Assay (metals basis)
Chemical Name or Material Silver perchlorate monohydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'