missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver perchlorate monohydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Description
Silver perchlorate can be used in the manufacturing of explosives. It is generally used as a source of silver ions during silver nanoparticles synthesis. It is used as a catalyst in organic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 485°C (decomposition) |
Color | White |
Physical Form | Crystalline |
Quantity | 250 g |
UN Number | UN1481 |
Merck Index | 14,8523 |
Solubility Information | Soluble in water and organic solvents. |
Formula Weight | 225.34 (207.32 Anhydrous) |
Percent Purity | 99.9% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'