missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver perchlorate hydrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Color | White |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 5 g |
Assay Percent Range | 98.5% min. (on dry substance) (Argentometry) |
Linear Formula | AgClO4·xH2O |
Merck Index | 10, 8358 |
Solubility Information | Solubility in water: 557g/100mL. Other solubilities: soluble in many organic solvents |
IUPAC Name | silver;perchlorate;hydrate |
Formula Weight | 207.32 |
Percent Purity | 99% |
Show More |
Safety and Handling
Product Identifier
- Silver perchlorate hydrate
- Danger
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'