missing translation for 'onlineSavingsMsg'
Learn More

Silver perchlorate, anhydrous, Thermo Scientific Chemicals

Catalog Number p-4407820
Click to view available options
Quantity:
5 g
25 g
100 g
7783-93-9
AgClO4
207.314
MFCD00003400
YDHABVNRCBNRNZ-UHFFFAOYSA-M
silver perchlorate, unii-sv4x1u113o, silver 1+ ion perchlorate, perchloric acid, silver 1+ salt, silver 1+ salt, ag.clo4, perchloric acid, silver 1+ salt 1:1, silver 1+ perchlorate ion, silver i perchloratehydrate, silver i perchlorate, anhydrous
24562
silver;perchlorate
[O-]Cl(=O)(=O)=O.[Ag+]
7783-93-9
AgClO4
207.314
MFCD00003400
YDHABVNRCBNRNZ-UHFFFAOYSA-M
silver perchlorate, unii-sv4x1u113o, silver 1+ ion perchlorate, perchloric acid, silver 1+ salt, silver 1+ salt, ag.clo4, perchloric acid, silver 1+ salt 1:1, silver 1+ perchlorate ion, silver i perchloratehydrate, silver i perchlorate, anhydrous
24562
silver;perchlorate
[O-]Cl(=O)(=O)=O.[Ag+]

Light Sensitive & Hygroscopic

Silver perchlorate is used as a catalyst in organic chemistry. It is an effective reagent for replacing halides ligands with perchlorate. It is also used in the manufacturing of explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 485°C (decomposition)
Color White
Physical Form Crystals
Quantity 5 g
Solubility Information Soluble in water and many organic solvents (benzene,toluene,pyridine)Soluble in water,benzene and toluene.
Formula Weight 207.32
Sensitivity Hygroscopic; Light sensitive
Appearance White crystals
Density 2.806 g/cm3 at 20°C
Chemical Name or Material Silver perchlorate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'