missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrite, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407819
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7783-99-5
AgNO2
153.873
MFCD00003413
KKKDGYXNGYJJRX-UHFFFAOYSA-M
silver nitrite, unii-t3mz57ogif, t3mz57ogif, silbernitrit, nitrous acid, silver 1+ salt, silver i nitrite, silvernitrite, acmc-20ajq7, ag.no2, silver nitrite 10g
160904
silver;nitrite
N(=O)[O-].[Ag+]
This item is not returnable. View return policy
7783-99-5
AgNO2
153.873
MFCD00003413
KKKDGYXNGYJJRX-UHFFFAOYSA-M
silver nitrite, unii-t3mz57ogif, t3mz57ogif, silbernitrit, nitrous acid, silver 1+ salt, silver i nitrite, silvernitrite, acmc-20ajq7, ag.no2, silver nitrite 10g
160904
silver;nitrite
N(=O)[O-].[Ag+]

Silver nitrite is used in the silver plating process, hair dye, inks, permanent marker pens and determination of chloride ions in water and drilling fluids. It is used as Tollen’s reagent and used in the formation of aniline and nitro compounds in victor mayer reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 140°C (decomposition)
Color White
Physical Form Crystalline
Quantity 10 g
Assay Percent Range 99% (metals basis)
Solubility Information Soluble in hot water.Insoluble in ethanol.
Formula Weight 153.88
Odor Odorless
Sensitivity Light sensitive
Appearance Light yellow crystals
Density 4.45 g/cm3
Chemical Name or Material Silver nitrite
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'