missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver nitrate solution, Volumetric, Reag. Ph. Eur., 0.1 M AgNO3 (0.1N), Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
6 x 1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Quantity | 5 L |
UN Number | UN3082 |
IUPAC Name | silver(1+) nitrate |
Formula Weight | 169.87g/mol |
Concentration or Composition (by Analyte or Components) | 0.1M |
Chemical Name or Material | Silver nitrate solution |
Safety and Handling
Product Identifier
- Silver nitrate solution
- Warning
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P234-Keep only in original container.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Silver nitrate solution
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'