missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate solution, Volumetric, 0.0282 M AgNO3 (1 ml = 1 mg Cl), Honeywell Fluka™

Product Code. 15665020 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
This item is not returnable. View return policy

Product Code. 15665020

Brand: Honeywell-Fluka 353776X1L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric, 0.0282 M AgNO3 (1 ml = 1 mg Cl)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8, 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 100°C
CAS Min % 99.0000%
CAS Max % 100.0000%, <1.0000%
Quantity 6 x 1 L
Linear Formula AgNO3
IUPAC Name silver;nitrate
Formula Weight 169.87
Concentration or Composition (by Analyte or Components) 0.0282 M AgNO3 (1 ml = 1 mg Cl)
Packaging Glass Bottle
Density 1.000 g/cm3
Chemical Name or Material Silver nitrate solution, Volumetric, 0.0282 M AgNO3 (1 ml = 1 mg Cl)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.