Learn More
Silver Nitrate R2 solution EU Pharmacopoeia, Fisher Chemical™
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Brand: Fisher Chemical RP 189 1000
Code : 04
Additional Details : CAS Number : 7761-88-8 Weight : 1.00000kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Specifications
1000 mL | |
EU Ph Ref: 1078302 | |
Eu. Ph. | |
Glass Bottle |
Safety and Handling
- silver nitrate solution R2
Signal Word
- Warning
Hazard Category
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : silver nitrate