missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, puriss. p.a. ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 25 g |
Linear Formula | AgNO3 |
UN Number | 1493 |
Formula Weight | 169.87g/mol |
Percent Purity | ≥99.8% |
Grade | ACS/puriss. p.a. |
Density | Vapor Density: 5.8 (vs air) |
Chemical Name or Material | Silver Nitrate, Reag. Ph. Eur. reag. ISO |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'