missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, puriss. p.a. ACS Reagent, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
25g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 250 g |
| Linear Formula | AgNO3 |
| UN Number | 1493 |
| Formula Weight | 169.87g/mol |
| Percent Purity | ≥99.8% |
| Grade | ACS/puriss. p.a. |
| Density | Vapor Density: 5.8 (vs air) |
| Chemical Name or Material | Silver Nitrate, Reag. Ph. Eur. reag. ISO |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction