missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, Certified AR for Analysis, Fisher Chemical™
938.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
Description
AgNO3, CAS Number-7761-88-8, silbernitrat, silver i nitrate, silver mononitrate, nitrate d'argent, silver nitrate, nitric acid silver i salt, silvernitrate, argenti nitras, argerol, lunar caustic, 500g, 444 deg.C, CHEBI:32130, White, 169.87g/mol, SQGYOTSLMSWVJD-UHFFFAOYSA-N, silver;nitrate
Specifications
Boiling Point | 444°C |
Melting Point | 212°C |
Color | White |
pH | 5.4 to 6.4 |
Physical Form | Solid |
Quantity | 250 g |
IUPAC Name | silver(1+) nitrate |
Formula Weight | 169.87g/mol |
Packaging | Glass Jar |
Safety and Handling
Product Identifier
- Silver nitrate
- Danger
- Oxidising solid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'