missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, Analytical Volumetric, 0.1M, Reagecon™
BIOPHARMA

Catalog Number p-8014530
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Determine concentrations using silver nitrate as the titrant.  Reagecon™ Silver Nitrate, Analytical Volumetric, 0.1M is perfect for titration of halides, anions that behave like halides, mercaptans and a variety of organic materials.

strong>Precipitation Titrations
  • This analytical methodology is based on reactions that yield compounds of limited solubility.
  • There is not a very wide range of precipitating agents that can be used gainfully in titrometry and silver nitrate is by far the most important.
  • These titrations, (also called argentometric titration) is where silver nitrate is used as the titrant.
  • Silver nitrate can be used for determination of halides (Cl-, I-, Br-) and anions that behave like halides (SCN-, CN-, CNO-).
  • It can also be used for determination of Mercaptans and organic materials that include Fatty Acids.
  • Indicators typically used for precipitation titrations include sodium chromate, fluorescein and iron (III).
  • A wide range of standardised silver nitrate titrants are available, some of which are standardised to specifically give a one to one equivalence with sodium chloride in various food stuffs.
Quantity 1 L
compliance-icons
Product Identifier
  • SILVER NITRATE 0.1N (0.1M)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : silver nitrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'