Learn More
Alfa Aesar™ Silver nitrate, Acculute Standard Volumetric Solution, Final Concentration 0.282N
Brand:  Alfa Aesar™ 088615.LQ
Code : NEW
Additional Details : CAS Number : 7761-88-8 Weight : 0.01000kg Transport : UN number : 3093 Chem class : 1 Pack group : II
Quantity 1 unit
 View more versions of this product
Product Code. 11349319
  • £264.60 / 1 unit
Estimated Shipment date
from Supplier 22-05-2018
Add to basket
Specifications
Chemical Identifiers
Silver nitrate Acculute Standard Volumetric Solution
Final Concentration 0.282N
169.87
UN3093
7761-88-8
AgNO3
MFCD00003414
Specifications
Silver nitrate Acculute Standard Volumetric Solution
7761-88-8
AgNO3
MFCD00003414
Colorless
Light Sensitive
Final Concentration 0.282N
Liquid
169.87
UN3093
1 unit