missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, 99.85%, for analysis, Thermo Scientific Chemicals

Catalog Number 10503951
30.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
[Ag+].[O-][N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Assay Percent Range 99.85%
Linear Formula AgNO3
Merck Index 15, 8657
IUPAC Name silver;nitrate
Formula Weight 169.87
Percent Purity ≥99.80%
Grade Analytical
Loss on Drying 0.1% (1g, 70°C, 2 hrs) max.
Packaging Glass bottle
Decomposition Information >444°C
Chemical Name or Material Silver nitrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'