missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver nanoparticles, 40nm, supplied in 2mM citrate, conjugated to Streptavidin

Brand:  Alfa Aesar™ J67442.XF

UNSPSC : 12352200

Code : NEW

Additional Details : Weight : 0.00001kg

Quantity 1mL

 View more versions of this product

Product Code. 15464635

  • £161.12 / 1mL


Estimated Shipment date
from Supplier 11-08-2017
Add to basket

Specifications

Description

Applications
Silver nanoparticles applications include molecular diagnostics and photonic devices, which take advantage of the novel optical properties of these nanomaterials. An increasingly common application is the use of silver nanoparticles for antimicrobial coatings, and many textiles, keyboards, wound dressings, and biomedical devices now contain silver nanoparticles that continuously release a low level of silver ions to provide protection against bacteria.

Solubility
Fully miscible in water.

Notes
Protect from heat, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark and keep it refrigerated.
Description & Specifications

Chemical Identifiers

Silver nanoparticles
supplied in 2mM citrate, conjugated to Streptavidin
MFCD00003397

Specifications

Silver nanoparticles
supplied in 2mM citrate, conjugated to Streptavidin
MFCD00003397
40nm
1mL
Liquid
Odorless
Yellow
Documentation

Technical Documents