missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver nanoparticles, 20nm, supplied in 2mM citrate, conjugated to Streptavidin

Brand:  Alfa Aesar™ J67221.XF

Code : NEW

Additional Details : Weight : 0.00001kg

Quantity 1mL

Product Code. 15494555

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Silver nanoparticles
supplied in 2mM citrate, conjugated to Streptavidin
Liquid
20nm
1mL
7440-22-4
MFCD00003397
Yellow
Videos
SDS