missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver nanoparticles, 100nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 490nm absorption, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
Description
An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range. Readily binds proteins and other ligands for conjugate and sensor development.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range
- Readily binds proteins and other ligands for conjugate and sensor development
Specifications
Color | Yellow |
Quantity | 100 mL |
Odor | Odorless |
Particle Size | 100nm |
Sensitivity | Light sensitive |
Form | Liquid |
Chemical Name or Material | Silver nanoparticles, 0.02mg/mL, supplied in 2mM sodium citrate, 490nm absorption |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'