missing translation for 'onlineSavingsMsg'
Learn More

Silver molybdenum oxide, 99%, Thermo Scientific Chemicals

Catalog Number p-7050889
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy
13765-74-7
MFCD00053384
Silver(I) molybdate
This item is not returnable. View return policy
13765-74-7
MFCD00053384
Silver(I) molybdate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 483°C
Color Beige
Physical Form Powder
Quantity 0.25 g
Formula Weight 375.68
Percent Purity 99%
Odor Odorless
Sensitivity Light sensitive
Density 6.18 g/mL
Chemical Name or Material Silver molybdenum oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'