missing translation for 'onlineSavingsMsg'
Learn More

Silver methanesulfonate, Thermo Scientific Chemicals

Catalog Number p-4824932
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
2386-52-9
CH3AgO3S
202.96
MFCD00064795
MLKQJVFHEUORBO-UHFFFAOYSA-M
silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
159658
silver;methanesulfonate
CS(=O)(=O)[O-].[Ag+]
This item is not returnable. View return policy
2386-52-9
CH3AgO3S
202.96
MFCD00064795
MLKQJVFHEUORBO-UHFFFAOYSA-M
silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
159658
silver;methanesulfonate
CS(=O)(=O)[O-].[Ag+]

Silver methanesulfonate is used as a catalyst for heterocyclization reactions. Further, it is used in the synthesis of unsaturated ketones and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 10 g
Solubility Information Soluble in water,alcohol,ether and benzene. Slightly soluble in toluene,o-chlorotoluene and ethyl disulfide. Insoluble in hexane and methylcyclopentane.
Formula Weight 202.97
Odor Odorless
Sensitivity Light sensitive
Appearance White powder
Chemical Name or Material Silver methanesulfonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'