missing translation for 'onlineSavingsMsg'
Learn More

Silver lactate, 96%, Thermo Scientific Chemicals

Catalog Number 11974351
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
15768-18-
C3H5O3Ag
196.94
LMEWRZSPCQHBOB-UHFFFAOYSA-M
silver lactate, silver lactate monohydrate, lactato-o1,o2 silver, lactic acid, silver salt, silver, 2-hydroxypropanoato-o1,o2, propanoic acid, 2-hydroxy-, silver complex, silver, 2-hydroxy-kappao propanoato-kappao
91824762
silver;2-hydroxypropanoate
CC(C(=O)[O-])O.[Ag+]
This item is not returnable. View return policy
15768-18-
C3H5O3Ag
196.94
LMEWRZSPCQHBOB-UHFFFAOYSA-M
silver lactate, silver lactate monohydrate, lactato-o1,o2 silver, lactic acid, silver salt, silver, 2-hydroxypropanoato-o1,o2, propanoic acid, 2-hydroxy-, silver complex, silver, 2-hydroxy-kappao propanoato-kappao
91824762
silver;2-hydroxypropanoate
CC(C(=O)[O-])O.[Ag+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 120.0°C to 122.0°C
Color Brown-Gray to Gray
Physical Form Powder
Quantity 50 g
Assay Percent Range 95% min. (Argentometry)
Linear Formula CH3CH(OH)CO2Ag
Merck Index 14,8517
Solubility Information Solubility in water: 77g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 196.94
Percent Purity 96%
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material Silver lactate
Show More Show Less
compliance-icons
Product Identifier
  • Silver lactate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'