missing translation for 'onlineSavingsMsg'
Learn More

Silver lactate, Thermo Scientific Chemicals

Catalog Number p-7049743
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
15768-18-0
C3H5AgO3
196.938
MFCD00043279
LMEWRZSPCQHBOB-UHFFFAOYSA-M
silver lactate, silver lactate monohydrate, lactato-o1,o2 silver, lactic acid, silver salt, silver, 2-hydroxypropanoato-o1,o2, propanoic acid, 2-hydroxy-, silver complex, silver, 2-hydroxy-kappao propanoato-kappao
91824762
silver;2-hydroxypropanoate
CC(C(=O)[O-])O.[Ag+]
This item is not returnable. View return policy
15768-18-0
C3H5AgO3
196.938
MFCD00043279
LMEWRZSPCQHBOB-UHFFFAOYSA-M
silver lactate, silver lactate monohydrate, lactato-o1,o2 silver, lactic acid, silver salt, silver, 2-hydroxypropanoato-o1,o2, propanoic acid, 2-hydroxy-, silver complex, silver, 2-hydroxy-kappao propanoato-kappao
91824762
silver;2-hydroxypropanoate
CC(C(=O)[O-])O.[Ag+]

Silver lactate is used as a surgical antiseptic. It is also used as an antimicrobial agent in the external applications. Further, it is used in the treatment of full thickness thermal wounds. In addition to this, it is employed as a hypodermic injection in erysipelas

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 120°C to 122°C
Color White to Gray
Physical Form Powder
Quantity 10 g
Linear Formula CH3CH(OH)CO2Ag
UN Number UN3077
Merck Index 14,8517
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 196.94
Odor Odorless
Sensitivity Light sensitive
Chemical Name or Material Silver lactate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'