missing translation for 'onlineSavingsMsg'
Learn More

Silver iodate, 98%, Thermo Scientific Chemicals

Catalog Number p-4407812
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7783-97-3
AgIO3
282.77
MFCD00014150
YSVXTGDPTJIEIX-UHFFFAOYSA-M
silver iodate, unii-5p7ox4vt1f, silver 1+ ion iodate, 5p7ox4vt1f, silberjodat, silver 1+ iodate, acmc-20ajqa, iodic acid hio3 , silver 1+ salt, ag.io3, silver iodate 25g
165642
silver;iodate
[O-]I(=O)=O.[Ag+]
This item is not returnable. View return policy
7783-97-3
AgIO3
282.77
MFCD00014150
YSVXTGDPTJIEIX-UHFFFAOYSA-M
silver iodate, unii-5p7ox4vt1f, silver 1+ ion iodate, 5p7ox4vt1f, silberjodat, silver 1+ iodate, acmc-20ajqa, iodic acid hio3 , silver 1+ salt, ag.io3, silver iodate 25g
165642
silver;iodate
[O-]I(=O)=O.[Ag+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >200°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Solubility Information Practically insoluble in water. Soluble in aqueous ammonia
Formula Weight 282.77
Odor Odorless
Sensitivity Light sensitive
Appearance White Powder
Density 5.53 g/cm3 at 20°C
Chemical Name or Material Silver iodate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'