missing translation for 'onlineSavingsMsg'
Learn More

Silver(II) oxide, 85.5-88.0% Ag, ACROS Organics™

Brand:  Acros Organics 207850500

UNSPSC : 12352300

Code : 29

Additional Details : CAS Number : 1301-96-8 Weight : 0.10000kg Transport : UN number : 3085 Chem class : 1a Pack group : II


Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10490671

  • £140.20 / 50g


Estimated Shipment
01-09-2017
Add to basket


Specifications

Chemical Identifiers

Silver(II) oxide
1301-96-8
AgO
123.87
85.5-88.0% Ag
85.5 to 88% (AG)
AgO
MFCD00044285

Specifications

Silver(II) oxide
1301-96-8
85.5 to 88% (AG)
AgO
MFCD00044285
15, 8661
Gray to black
85.5-88.0% Ag
Powder
AgO
123.87
02,369; 04,431; 06,518; 07,322; 09,412; 10,352; 11,469
Glass bottle
50g
Safety and Handling

Safety and Handling

Product Identifier
  • Silver(II) oxide

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Documentation

Technical Documents