missing translation for 'onlineSavingsMsg'
Learn More

Silver(II) fluoride, 98+%, Thermo Scientific Chemicals

Catalog Number p-4824944
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
7783-95-1
AgF2
MFCD00003411
Silver difluoride
This item is not returnable. View return policy
7783-95-1
AgF2
MFCD00003411
Silver difluoride

Moisture and Light Sensitive

Silver(II) fluoride is used as a fluorinating and oxidizing agent. It is also used in the preparation of organic perfluorocompounds and in the fluorination of aromatic compounds. In addition, it is also used in pharmacy and electroplating industry. It serves as an intermediate in the catalysis of gaseous reactions with fluorine by silver. It is utilized in the oxidation of xenon to xenon difluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 690°C
Color Brown
Physical Form Crystalline
Quantity 2 g
Assay Percent Range 98+%
Solubility Information Soluble in hexane,chloroform,chlorofluorocarbon and hydrogen fluoride.
Formula Weight 145.87
Sensitivity Light and moisture sensitive
Appearance Brown crystals
Density 4.6
Chemical Name or Material Silver(II) fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'