missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) fluoride, 99+%, ACROS Organics™

£60.50 - £212.80

Chemical Identifiers

CAS 7775-41-9
Molecular Formula AgF
Molecular Weight (g/mol) 126.87
MDL Number MFCD00003410
InChI Key REYHXKZHIMGNSE-UHFFFAOYSA-M
Synonym silver fluoride, silver 1+ fluoride, silver 1+ ion fluoride, argentous fluoride, ag-f, silverfluoride, silver salt, fluoride, silver fluoride,, silver i fluoride 5g
PubChem CID 165912
ChEBI CHEBI:30340
IUPAC Name silver;fluoride
SMILES [F-].[Ag+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10022630 SDS View Documents Acros Organics
210450050
5g Plastic bottle £60.50
5g View alternative products
Save Up to 
»
EU Stock 8 Please to purchase this item. Need a web account? Register with us today!
10531081 SDS View Documents Acros Organics
210450250
25g Plastic bottle £212.80
25g View alternative products
Save Up to 
»
EU Stock 7 Please to purchase this item. Need a web account? Register with us today!
Description

Description

C-X bond formation reagent
Specifications

Chemical Identifiers

7775-41-9
126.87
REYHXKZHIMGNSE-UHFFFAOYSA-M
165912
silver;fluoride
AgF
MFCD00003410
silver fluoride, silver 1+ fluoride, silver 1+ ion fluoride, argentous fluoride, ag-f, silverfluoride, silver salt, fluoride, silver fluoride,, silver i fluoride 5g
CHEBI:30340
[F-].[Ag+]

Specifications

Silver(I) fluoride
7775-41-9
AgF
MFCD00003410
15, 8653
Solubility in water: in water: 182g/100mL (15.5°C)
[F-].[Ag+]
126.87
CHEBI:30340
Powder
5.8500g/mL
Solubility in water: in water: 182 g/100mL (15.5°C).
1150.0°C
435.0°C
99+%
99% min
AgF
02,364; 05,581; 06,514; 15,287
silver fluoride, silver 1+ fluoride, silver 1+ ion fluoride, argentous fluoride, ag-f, silverfluoride, silver salt, fluoride, silver fluoride,, silver i fluoride 5g
REYHXKZHIMGNSE-UHFFFAOYSA-M
silver;fluoride
165912
126.87
99+%
Plastic bottle
5.85
Brown-Rust to Brown-Yellow
5g
Safety and Handling

Safety and Handling

Product Identifier
  • Silver(I) fluoride

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.