missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver(I) citrate hydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Silver(I) citrate hydrate can be very attractive for the future biomedical or pharmaceutical/therapeutic applications. It can be used as an antimicrobial agent and enhancers of antioxidants and anticancer agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 170°C (decomposition) |
Quantity | 25 g |
Sensitivity | Light sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 512.70 (Anhydrous) |
Physical Form | Powder |
Chemical Name or Material | Silver(I) citrate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'