missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver hydrogen fluoride, 99% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Silver hydrogen fluoride facilitates the photochemical conversion of carboxylic acids to alkyl fluorides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Physical Form | Crystalline |
Quantity | 10 g |
Linear Formula | AgHF2 |
UN Number | UN1740 |
Solubility Information | Soluble in water. |
Formula Weight | 146.87 |
Percent Purity | ≤99% |
Sensitivity | Hygroscopic; Light sensitive |
Chemical Name or Material | Silver hydrogen fluoride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'