missing translation for 'onlineSavingsMsg'
Learn More

Silver hexafluorophosphate, 98%, Thermo Scientific Chemicals

Catalog Number p-4412860
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
26042-63-7
AgF6P
252.832
MFCD00003415
SCQBROMTFBBDHF-UHFFFAOYSA-N
silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
168464
silver;hexafluorophosphate
F[P-](F)(F)(F)(F)F.[Ag+]
This item is not returnable. View return policy
26042-63-7
AgF6P
252.832
MFCD00003415
SCQBROMTFBBDHF-UHFFFAOYSA-N
silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
168464
silver;hexafluorophosphate
F[P-](F)(F)(F)(F)F.[Ag+]

It is commonly used as an oxidant forming silver metal. It is often used as a reagent in inorganic and organometallic chemistry. It is used in the preparation of engineering novel materials as well as in preparation of supra molecular Ag(I) complexes. It is used in the development of new catalysts, replacing halide ligands with the weakly coordinating hexafluorophosphate anion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 102°C (decomposition)
Color Brown to Yellow
Physical Form Powder
Quantity 1 g
Assay Percent Range 98%
Solubility Information Soluble in organic solvents such as benzene,toluene,and acetonitrile.
Formula Weight 252.83
Odor Odorless
Sensitivity Light sensitive
Appearance White powder
Chemical Name or Material Silver hexafluorophosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'