missing translation for 'onlineSavingsMsg'
Learn More

Silver hexafluorophosphate, 98%, -6 mesh, Thermo Scientific Chemicals

Catalog Number p-3735355
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
26042-63-7
AgF6P
252.83
MFCD00003415
SCQBROMTFBBDHF-UHFFFAOYSA-N
silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
168464
silver;hexafluorophosphate
F[P-](F)(F)(F)(F)F.[Ag+]
This item is not returnable. View return policy
26042-63-7
AgF6P
252.83
MFCD00003415
SCQBROMTFBBDHF-UHFFFAOYSA-N
silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
168464
silver;hexafluorophosphate
F[P-](F)(F)(F)(F)F.[Ag+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Beige-Gray to White
Physical Form Powder
Quantity 1 g
Linear Formula AgPF6
Solubility Information Solubility in water: soluble
Formula Weight 252.83
Percent Purity 98%
Packaging Plastic bottle
Chemical Name or Material Silver hexafluorophosphate
compliance-icons
Product Identifier
  • Silver hexafluorophosphate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'