missing translation for 'onlineSavingsMsg'
Learn More

Silver hexafluoroarsenate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7048833
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
12005-82-2
AgAsF6
296.78
MFCD00014142
UNSKNPXMHZPWPB-UHFFFAOYSA-N
silver hexafluoroarsenate, silver hexafluoroarsenate v, silver hexafluoroarsoranuide, acmc-1c478, silver hexakis fluoranyl arsenic 1-, silver 1+ hexafluoro-??-arsanuide, silver 1+ ion hexafluoro-??-arsanuide
11077325
silver;hexafluoroarsenic(1-)
F[As-](F)(F)(F)(F)F.[Ag+]
This item is not returnable. View return policy
12005-82-2
AgAsF6
296.78
MFCD00014142
UNSKNPXMHZPWPB-UHFFFAOYSA-N
silver hexafluoroarsenate, silver hexafluoroarsenate v, silver hexafluoroarsoranuide, acmc-1c478, silver hexakis fluoranyl arsenic 1-, silver 1+ hexafluoro-??-arsanuide, silver 1+ ion hexafluoro-??-arsanuide
11077325
silver;hexafluoroarsenic(1-)
F[As-](F)(F)(F)(F)F.[Ag+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 2 g
UN Number UN1557
Formula Weight 296.78
Percent Purity ≥98%
Odor Odorless
Sensitivity Moisture Sensitive
Chemical Name or Material Silver hexafluoroarsenate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'