missing translation for 'onlineSavingsMsg'
Learn More

Silver gauze, 80 mesh woven from 0.115mm (0.0045in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4824931
34.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 x 25 mm
75 x 75 mm
150 x 150 mm
300 x 300 mm
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

Silver gauze is used for filtration. It is also used in electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver
Appearance Silver-colored guaze
Assay Percent Range 99.9% (Metals basis)
Quantity 25 x 25 mm
Solubility Information Insoluble in water.
Mesh Size 80 Mesh
Odor Characteristic
Form Wire Cloth
Chemical Name or Material Silver

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'