missing translation for 'onlineSavingsMsg'
Learn More

Silver foil, 99.998% (metals basis), Alfa Aesar™

Form: approx. 2.6g/50x50mm; Temper: soft

Brand:  Alfa Aesar 042317.GH

Code : NEW

Additional Details : CAS Number : 7440-22-4 Weight : 0.01000kg

Quantity 100 x 100mm

 View more versions of this product

Product Code. 11304378

  • £112.40 / Each


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

Silver
7440-22-4
99.998% (Metals basis)
Annealed
Foil, 0.1mm (0.004 in.) thick
MFCD00003397

Specifications

Silver
Annealed
Silver-colored foil
99.998% (Metals basis)
Characteristic
100 x 100mm
7440-22-4
Foil, 0.1mm (0.004 in.) thick
MFCD00003397
Silver
Videos
SDS