missing translation for 'onlineSavingsMsg'
Learn More

Silver foil, 1.0mm (0.04in) thick, Premion™, 99.9985% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4824930
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
This item is not returnable. View return policy
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
This item is not returnable. View return policy
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

Form: Approx. 6.6g/25 x 25mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver
Appearance Silver-colored foil
Assay Percent Range 99.9985% (Metals basis)
Quantity 25 x 25 mm
Odor Characteristic
Form Foil
Chemical Name or Material Silver

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'