missing translation for 'onlineSavingsMsg'
Learn More

Silver foil, 0.025mm (0.001in) thick, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4824926
Click to view available options
Quantity:
150 x 150 mm
150 x 600 mm
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

Form: Approx. 5.9g/150 x 150mm; Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver
Appearance Silver-colored foil
Assay Percent Range 99.95% (Metals basis)
Quantity 150 x 150 mm
Odor Characteristic
Form Foil
Chemical Name or Material Silver, Annealed

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'