missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver flake, 80% <20 micron, 99.9% (metals basis)

Brand:  Alfa Aesar™ 014639.30

Code : NEW

Additional Details : CAS Number : 7440-22-4 Weight : 0.25000kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Quantity 250g

 View more versions of this product

Product Code. 11316958

  • £606.20 / 250g


Estimated Shipment date
from Supplier 25-06-2018
Add to basket

Specifications

Chemical Identifiers

Silver flake
7440-22-4
MFCD00003397
UN3077

Specifications

Silver flake
250g
Flakes
(metals basis)
UN3077
80% <20 Micron
Scott density: 18g/cm3 to 28g/in3
7440-22-4
99.9%
MFCD00003397
Characteristic
1.0 to 2.2m2/g
Videos
SDS