missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver cyclohexanebutyrate

Brand:  Alfa Aesar™ 043708.06

Code : NEW

Additional Details : CAS Number : 62638-04-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11350308

  • £43.67 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Silver cyclohexanebutyrate
C10H17AgO2
MFCD00036396
62638-04-4
277.11
Cyclohexanebutyric acid silver salt; 4-Cyclohexylbutyric acid silver salt

Specifications

Silver cyclohexanebutyrate
Powder
277.11
Cyclohexanebutyric acid silver salt; 4-Cyclohexylbutyric acid silver salt
≈255°C (dec.)
62638-04-4
C10H17AgO2
MFCD00036396
Light Sensitive
5g
Videos
SDS