missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver conductive coating

Brand:  Alfa Aesar™ 044937.10

Code : NEW

Additional Details : Weight : 0.01500kg Transport : UN number : 1133 Chem class : 2 Pack group : II

Quantity 15g

Product Code. 15462077

  • £95.80 / 15g


Estimated Shipment date
from Supplier 15-02-2018
Add to basket

Specifications

Specifications

Silver conductive coating
MFCD00003397
15g
Sheet resistance 0.015 ohms/square in./mil (25μm)
UN1133
Videos
SDS