missing translation for 'onlineSavingsMsg'
Learn More

Silver chloride, ultra dry, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050143
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7783-90-6
AgCl
143.32
MFCD00003399
HKZLPVFGJNLROG-UHFFFAOYSA-M
silver(1+) chloride
[Cl-].[Ag+]
This item is not returnable. View return policy
7783-90-6
AgCl
143.32
MFCD00003399
HKZLPVFGJNLROG-UHFFFAOYSA-M
silver(1+) chloride
[Cl-].[Ag+]

It is used in silver plating, electrode, photographic paper, in making antiseptic silver preparations. It is used to make photographic film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 1550°C
Melting Point 455°C
Color White
Physical Form Beads
Quantity 1 g
UN Number UN3077
Merck Index 14,8509
Solubility Information Soluble in alkali cyanides,thiosulfates,ammonium carbonates,ammonium chloride,mercuric nitrate,and silver nitrate. Insoluble in water.
Formula Weight 143.32
Percent Purity 99.997%
Odor Odorless
Packaging Ampule
Refractive Index 2.071
Sensitivity Light sensitive
Density 5.56 g/mL
Assay (metals basis)
Chemical Name or Material Silver chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'