missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver chloride crystal optic disc, 32mm x 3mm (drilled), polished both sides

Brand:  Alfa Aesar™ 041745.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15416395

  • £683.38 / Each


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Specifications

Silver chloride crystal optic disc
32mm x 3mm (drilled), Polished both sides
1 each
Videos
SDS