missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver chloride, 4.9% in H{2}O, nanoparticle dispersion, <200nm APS Number Weighted, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Used as pharmaceutical intermediate and in pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Density | 1.038 g/mL |
UN Number | UN3082 |
Quantity | 100 g |
Merck Index | 14,8509 |
Solubility Information | Dispersible in water. Very slightly soluble in cold water,soluble in solutions of alkali cyanides,thiosulfates,ammonium carbonates. Appreciably soluble in concentrated aqueous solutions of ammonium chloride,mercuric nitrate,and silver nitrate. |
Formula Weight | 143.32 |
Concentration or Composition (by Analyte or Components) | 4.9% in H2O |
Odor | Odorless |
Particle Size | <200nm APS Number Weighted |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'