Learn More
Thermo Scientific Alfa Aesar SILVER CASTING GRAIN 1.6- 6.35MM 3N% 1KG
Brand: Thermo Scientific Alfa Aesar 040511.A1
SILVER CASTING GRAIN 1.6- 6.35MM 3N% 1KG
Code : NEW
Additional Details : CAS Number : 7440-22-4 Weight : 1.00000kg
Brand: Thermo Scientific Alfa Aesar 040511.A1
SILVER CASTING GRAIN 1.6- 6.35MM 3N% 1KG
Code : NEW
Additional Details : CAS Number : 7440-22-4 Weight : 1.00000kg