missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver carbonate on Celite, ≈0.7 mmole Ag2CO3/g reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
15 mmol
75 mmol
Description
Silver carbonate is used in production of silver powder. It is used in microelectronics. Silver carbonate is used as a reagent in organic synthesis. It is employed in Fetizon oxidation, silver carbonate on celite acts as an oxidizing agent to form lactones from diols. It is also used in used in Koenigs-Knorr glycosylation. It is also employed to convert alkyl bromides into alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 15 mmol |
Solubility Information | Soluble in water (0.03mg/ml at 20°C),dilute HNO3,ammonia and ammonium hydroxide. Insoluble in alcohol. |
Odor | Odorless |
Sensitivity | Light sensitive |
Chemical Name or Material | Silver carbonate on Celite, ∼0.7 mmole Ag2CO3/g reagent |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'