missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver carbonate, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Silver carbonate is used in the production of silver powder in microelectronics, conducting polymers, and in tooth surface treatment to prevent cavities. It is also used as an antimicrobial agent for infection resistant surgical fabric materials. It is used in organic synthesis, and in biological specimen staining.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 218°C (decomposition) |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99.5% (metals basis) |
Solubility Information | Insoluble in water,acetone,acetate and alcohol. Freely soluble in dilute HNO3 and ammonia. |
Formula Weight | 275.75 |
Sensitivity | Light sensitive |
Density | 6.08 g/mL |
Chemical Name or Material | Silver carbonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'