missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver, brazing flux, white paste

Brand:  Alfa Aesar™ 041355.TB

Code : NEW

Additional Details : CAS Number : 7440-22-4 Weight : 0.11300kg

Quantity 113g

 View more versions of this product

Product Code. 11314068

  • £10.70 / 113g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

Silver, brazing flux
Active range: 540 to 980°
MFCD01868499

Specifications

Silver, brazing flux
Paste
White
Active range: 540 to 980°
MFCD01868499
113g
Videos
SDS