missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Silver, brazing alloy, flux covered, 1.6mm (0.06 in.) dia., 46cm (18 in.) long

Brand:  Thermo Scientific Alfa Aesar 041031.KJ

Silver, brazing alloy, flux covered, 1.6mm (0.06 in.) dia., 46cm (18 in.) long

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15436355

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.