missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver, brazing alloy, 1.6mm (0.06 in.) dia.

Brand:  Alfa Aesar™ 041029.10

Code : NEW

Additional Details : CAS Number : 7440-22-4 Weight : 0.01500kg

Quantity 15g

 View more versions of this product

Product Code. 11379437

  • £50.46 / 15g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

Silver, brazing Alloy
1.6mm (0.06 in.) dia.
MFCD00801120

Specifications

Ag:Cu:Zn:Sn; 56:22:17:5 wt%
15g
Nontoxic, Cadmium Free
Silver, brazing Alloy
1.6mm (0.06 in.) dia.
MFCD00801120
Videos
SDS