Learn More
Thermo Scientific Alfa Aesar Silver, brazing alloy, 1.6mm (0.06 in.) dia.
Brand: Thermo Scientific Alfa Aesar 041029.15
Silver, brazing alloy, 1.6mm (0.06 in.) dia.
Code : NEW
Additional Details : CAS Number : 7440-22-4 Weight : 0.03000kg