missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Silver bis(trifluoromethylsulfonyl)imide is used in the extraction of C5 - C8 linear alfa olefins using its silver salt to form room temperature ionic liquids. Further, it is utilized for the separation of olefins from paraffins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Melting Point | 249°C |
Color | Orange to White |
Physical Form | Solid |
Quantity | 1 g |
Linear Formula | (CF3SO2)2NAg |
UN Number | UN1759 |
Solubility Information | Soluble in water. |
Formula Weight | 388 |
Sensitivity | Light and moisture sensitive |
Chemical Name or Material | Silver bis (trifluoromethylsulfonyl)imide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'