missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver on alumina, 3%-4% Ag

Brand:  Alfa Aesar™ 045034.36

Code : NEW

Additional Details : Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 15482107

  • £528.60 / 500g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Silver on alumina
MFCD00003397

Specifications

Silver on alumina
MFCD00003397
500g
Spheres
3% to 4% Ag
2 to 4mm
Videos
SDS