missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver acetate, 99%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Beige-Gray to White |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99% |
Linear Formula | CH3CO2Ag |
Fieser | 01,1002; 03,362; 06,511; 08,440; 12,256; 13,285 |
Merck Index | 15,8644 |
Solubility Information | Solubility in water: 10.2g/L (20°C). Other solubilities: soluble in nitric acid |
Formula Weight | 166.91 |
Percent Purity | 99% |
Show More |
Safety and Handling
Product Identifier
- Silver acetate
- Warning
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Skin corrosion/irritation Category 2
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'